ContactUsLinkComponent
Syringe filters, Acrodisc® ion chromatography (IC)
Syringe filters, Acrodisc® ion chromatography (IC)
   Sustainable Options Available
Certificates
undefined
Syringe filters, Acrodisc® ion chromatography (IC)
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 514-4040
  • 514-4234
  • 516-7714
  • 514-4021
  • 514-4023
  • 516-7704
  • 516-7705
  • 514-4233
  • 514-4039
  • 516-7712
  • Type:
  • Syringe filter
  • Premium syringe filter (PSF)
  • Syringe filter
  • Syringe filter
  • Syringe filter
  • Syringe filter
  • Syringe filter
  • Premium syringe filter (PSF)
  • Syringe filter
  • Syringe filter
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Ø:
  • 25 mm
  • 25 mm
  • 25 mm
  • 13 mm
  • 13 mm
  • 13 mm
  • 13 mm
  • 25 mm
  • 25 mm
  • 25 mm
  • Pore size:
  • 0,45 µm
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,20 µm
  • Max. operating pressure:
  • -
  • 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C
  • -
  • 6,9 bar
  • 6,9 bar
  • 6,9 bar
  • 6,9 bar
  • 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C
  • -
  • -
  • Max. temperature:
  • -
  • 100 °C
  • -
  • 55 °C
  • 55 °C
  • 55 °C
  • 55 °C
  • 100 °C
  • -
  • -
  • Water flow rate:
  • -
  • 420 ml/min (2,1 bar)
  • -
  • 15 ml/min (1 bar)
  • 20 ml/min (1 bar)
  • 15 ml/min (1 bar)
  • 20 ml/min (1 bar)
  • 420 ml/min (2,1 bar)
  • -
  • -