ContactUsLinkComponent
Syringe filters, Minisart® NY
Syringe filters, Minisart® NY
Supplier:  Sartorius
Certificates
undefined
Syringe filters, Minisart® NY
Supplier:  Sartorius
Certificates
Specifications
  • Catalog No:
  • 514-0083
  • SART1776B-Q
  • 516-0681
  • SART17845-R
  • SART1776B-K
  • 514-0082
  • 516-0684
  • 514-0081
  • SART17846-R
  • SART1784B-K
  • 514-0080
  • SART1784B-Q
  • SART1784C-Q
  • SART1776C-Q
  • Pore size:
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,20 µm
  • 0,20 µm
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • 0,45 µm
  • 0,45 µm
  • Airflow rate:
  • 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
  • 20 ml/min with water at 1 bar (14,5 psi) | 40 ml/min with methanol at 1 bar (14,5 psi)
  • 40 ml/min with water at 1 bar (14,5 psi) | 110 ml/min with methanol at 1 bar (14,5 psi)
  • 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
  • 20 ml/min with water at 1 bar (14,5 psi) | 40 ml/min with methanol at 1 bar (14,5 psi)
  • 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
  • 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
  • 50 ml/min with water at 1 bar (14,5 psi) | 70 ml/min with methanol at 1 bar (14,5 psi)
  • 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
  • 50 ml/min with water at 1 bar (14,5 psi) | 70 ml/min with methanol at 1 bar (14,5 psi)
  • 50 ml/min with water at 1 bar (14,5 psi) | 70 ml/min with methanol at 1 bar (14,5 psi)
  • 50 ml/min with water at 1 bar (14,5 psi) | 70 ml/min with methanol at 1 bar (14,5 psi)
  • 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
  • 40 ml/min with water at 1 bar (14,5 psi) | 110 ml/min with methanol at 1 bar (14,5 psi)
  • Max. operating pressure:
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • Max. temperature:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)