Poly-lysine culture dishes, Corning® BioCoat™
Supplier: Corning
Certificates
About this item
PS, coated with poly-lysine.
Poly-D-lysine (PDL) and Poly-L-lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. Poly-lysine surface treatments support applications including attachment and spreading of a variety of cell lines; cell differentiation and neurite outgrowth; attachment of transfected cell lines; and survival of primary neurons in culture.
Specifications
- Catalog No:
- 734-0150
- 734-0301
- 734-0148
- 734-0302
- 734-0284
- 734-0149
- 734-0417
- 734-0178
- 734-0177
- 734-0285
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk