Tubing connectors, Y-shaped
Supplier: KARTELL
Certificates
About this item
PP, translucent.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Suitable for foodstuff
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine 'grip'. This will then give equivalent crest dimension for the particular connector concerned.
They meet with the Food and Drug regulations.90/128CEE 92/39CEE and FDA 21 CFR 177.1520.
Suitable for foodstuff.
Certifications: Conforms to Food and Drug regulations 90/128 EEC 92/39 EEC and FDA 21 CFR 177.1520.
Frequently Bought Together