ContactUsLinkComponent
Anion exchange HPLC columns, PRP-X600
Anion exchange HPLC columns, PRP-X600
Supplier:  HAMILTON BONADUZ
Certificates
undefined
Anion exchange HPLC columns, PRP-X600
Supplier:  HAMILTON BONADUZ
Specifications
  • Catalog No:
  • 554-1243
  • 554-1239
  • 554-0208
  • HAMI79220
  • 554-1244
  • Max. pressure:
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • pH range:
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • Temperature limit:
  • 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
  • 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
  • 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
  • 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
  • 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
Specifications
Related Information
Guard cartridge holder
Product Information