Orders LinkContactUsLinkComponent
HPLC columns, LiChrospher®
HPLC columns, LiChrospher®
Dodavatel:   Merck
HPLC columns, LiChrospher®
Dodavatel:  Merck
Certifikáty
HPLC columns, LiChrospher®
HPLC columns, LiChrospher®
84 Dostupných variant níže
Product Details & Documents
LiChrospher® is Merck's reliable and versatile, traditionally-produced, spherical silica. LiChrospher® is available with different modifications.
LiChrospher® Si 60 and Si 100 are versatile HPLC spherical silica sorbents for normal phase chromatography.
LiChrospher® CN has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. In addition, selective charged interactions makes it even more versatile.
LiChrospher® NH₂ possesses polar and hydrophobic properties and can be used for normal phase, reversed phase and ion exchange chromatography. Typical applications are the separations of carbohydrates by reversed phase chromatography or the separation of nucleotides by ion exhange chromatography.
LiChrospher® DIOL has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. LiChrospher® DIOL is also suitable for exclusion chromatography.
LiChrospher® RP-select B is a highly reproducible, traditionally-produced spherical silica gel with reversed phase properties. This sorbent is ideal for the separation of basic compounds, but can also be used with neutral and acidic samples.
LiChrospher® RP-8, RP-18 and RP-18 endcapped are highly reproducible and versatile traditionally-produced spherical silica gel sorbents with reversed-phase properties. They are ideal for the separation of neutral, acidic and weak basic compounds.
LiChrospher® WP 300 RP-18 is a highly selective and reliable HPLC column for the separation of peptides and low molecular weight proteins. LiChrospher® WP 300 RP-18 enables the tailing-free separation of basic compounds, and is suitable for the separation of tRNA.
LiChrospher® PAH is a highly efficient and selective HPLC column, especially designed for the high resolution separation of 16 PAH (polycyclic aromatic hydrocarbons) according to EPA 610 and 550, plus benzo(e)pyrene and perylene.
LiChroCART® cartridges (2, 3, 4, 4,6 mm I-Ø) require the use of manu-CART® cartridge holder 1.51486.0001
LiChroCART® cartridges (10×250 mm) require the use of manu-CART® cartridge holder 1.51419.0001
Guard cartridges have to be connected to the LiChroCART® cartridges using manu-CART® cartridge holder 1.51486.0001.
LiChrospher® RP-8, RP-18 and RP-18 endcapped are highly reproducible and versatile traditionally produced spherical silica gel sorbents with reversed phase properties. They are well suited for the separation of neutral, acidic and weak basic compounds.
Product Family Options
LiChrospher® LiChroCART® 3 mm ID cartridges
Informace o produktu
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Dostupnost
    Price
  • Analytical
    RP-select B
    60 Å
    5 µm
    3,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50158.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    3,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50155.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    3,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50159.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    3,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50154.0001
LiChrospher® LiChroCART® 4/4,6 mm ID cartridges
Informace o produktu
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Dostupnost
    Price
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,00 mm
    25 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 3 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50937.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,00 mm
    75 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 36 600,00 Kč
    36 600,00 Kč
    1 * 3 KS
       36 600,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50993.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 40 730,00 Kč
    40 730,00 Kč
    1 * 3 KS
       40 730,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50981.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50829.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 47 820,00 Kč
    47 820,00 Kč
    1 * 3 KS
       47 820,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50984.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50828.0001
  • Analytical
    NH₂
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 25 620,00 Kč
    25 620,00 Kč
    1 * 1 KS
       25 620,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50834.0001
  • Analytical
    Si
    60 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 320,00 Kč
    24 320,00 Kč
    1 * 1 KS
       24 320,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50830.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50832.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 25 620,00 Kč
    25 620,00 Kč
    1 * 1 KS
       25 620,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50833.0001
  • Analytical
    DIOL
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50836.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50837.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 030,00 Kč
    23 030,00 Kč
    1 * 1 KS
       23 030,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50838.0001
  • Analytical
    CN
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 25 620,00 Kč
    25 620,00 Kč
    1 * 1 KS
       25 620,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50892.0001
  • Analytical
    CN
    100 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 260,00 Kč
    23 260,00 Kč
    1 * 1 KS
       23 260,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50845.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,60 mm
    100 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 18 190,00 Kč
    18 190,00 Kč
    1 * 1 KS
       18 190,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50600.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 200,00 Kč
    22 200,00 Kč
    1 * 1 KS
       22 200,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50601.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50602.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    25 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 3 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50931.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    75 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 36 600,00 Kč
    36 600,00 Kč
    1 * 3 KS
       36 600,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50987.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 42 500,00 Kč
    42 500,00 Kč
    1 * 3 KS
       42 500,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50943.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 200,00 Kč
    22 200,00 Kč
    1 * 1 KS
       22 200,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50641.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 51 000,00 Kč
    51 000,00 Kč
    1 * 3 KS
       51 000,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50983.0001
  • Analytical
    RP-18
    100 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 260,00 Kč
    23 260,00 Kč
    1 * 1 KS
       23 260,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50843.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 200,00 Kč
    22 200,00 Kč
    1 * 1 KS
       22 200,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50638.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50639.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,00 mm
    25 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 3 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50936.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 200,00 Kč
    22 200,00 Kč
    1 * 1 KS
       22 200,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50635.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50636.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 51 000,00 Kč
    51 000,00 Kč
    1 * 3 KS
       51 000,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50995.0001
  • Analytical
    RP-18e
    100 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 260,00 Kč
    23 260,00 Kč
    1 * 1 KS
       23 260,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50848.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,60 mm
    100 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50603.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 200,00 Kč
    22 200,00 Kč
    1 * 1 KS
       22 200,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50604.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50605.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,00 mm
    25 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 3 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50930.0001
  • Analytical
    DIOL
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50826.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50827.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 3 KS
    1 * 3 KS - 48 760,00 Kč
    48 760,00 Kč
    1 * 3 KS
       48 760,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50982.0001
  • Analytical
    RP-8
    100 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 260,00 Kč
    23 260,00 Kč
    1 * 1 KS
       23 260,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50842.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50823.0001
  • Analytical
    NH₂
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50824.0001
  • Analytical
    CN
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50825.0001
  • Analytical
    Si
    60 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 18 890,00 Kč
    18 890,00 Kč
    1 * 1 KS
       18 890,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50820.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 310,00 Kč
    20 310,00 Kč
    1 * 1 KS
       20 310,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50822.0001
  • Analytical
    RP-8e
    100 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 260,00 Kč
    23 260,00 Kč
    1 * 1 KS
       23 260,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50847.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 440,00 Kč
    24 440,00 Kč
    1 * 1 KS
       24 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50642.0001
  • Analytical
    RP-select B
    60 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 21 840,00 Kč
    21 840,00 Kč
    1 * 1 KS
       21 840,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50742.0001
  • Analytical
    Si
    60 Å
    10 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 21 960,00 Kč
    21 960,00 Kč
    1 * 1 KS
       21 960,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50840.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 25 620,00 Kč
    25 620,00 Kč
    1 * 1 KS
       25 620,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50839.0001
LiChrospher® LiChroCART® 10 mm ID cartridges
Informace o produktu
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Dostupnost
    Price
  • Analytical
    RP-18e
    100 Å
    10 µm
    10,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 51 000,00 Kč
    51 000,00 Kč
    1 * 1 KS
       51 000,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50858.0001
LiChrospher® guard cartridges
Informace o produktu
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Dostupnost
    Price
  • Guard cartridge
    RP-select B
    60 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 880,00 Kč
    14 880,00 Kč
    1 * 10 KS
       14 880,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50963.0001
  • Guard cartridge
    Si
    60 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 290,00 Kč
    14 290,00 Kč
    1 * 10 KS
       14 290,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50955.0001
  • Guard cartridge
    CN
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 290,00 Kč
    14 290,00 Kč
    1 * 10 KS
       14 290,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50959.0001
  • Guard cartridge
    DIOL
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 170,00 Kč
    14 170,00 Kč
    1 * 10 KS
       14 170,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50960.0001
  • Guard cartridge
    NH₂
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 880,00 Kč
    14 880,00 Kč
    1 * 10 KS
       14 880,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50958.0001
  • Guard cartridge
    RP-18e
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 290,00 Kč
    14 290,00 Kč
    1 * 10 KS
       14 290,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50962.0001
  • Guard cartridge
    RP-18
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 880,00 Kč
    14 880,00 Kč
    1 * 10 KS
       14 880,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50957.0001
  • Guard cartridge
    RP-8e
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 290,00 Kč
    14 290,00 Kč
    1 * 10 KS
       14 290,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50961.0001
  • Guard cartridge
    RP-8
    100 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 290,00 Kč
    14 290,00 Kč
    1 * 10 KS
       14 290,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50956.0001
LiChrospher® Hibar® RT colums fixed with end-fittings
Informace o produktu
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Dostupnost
    Price
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,60 mm
    100 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 19 960,00 Kč
    19 960,00 Kč
    1 * 1 KS
       19 960,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50573.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 910,00 Kč
    24 910,00 Kč
    1 * 1 KS
       24 910,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50574.0001
  • Analytical
    RP-select B
    60 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50575.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    125 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 080,00 Kč
    22 080,00 Kč
    1 * 1 KS
       22 080,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50477.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50377.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,60 mm
    100 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 19 960,00 Kč
    19 960,00 Kč
    1 * 1 KS
       19 960,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50545.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 910,00 Kč
    24 910,00 Kč
    1 * 1 KS
       24 910,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50546.0001
  • Analytical
    RP-18
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50547.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 910,00 Kč
    24 910,00 Kč
    1 * 1 KS
       24 910,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50549.0001
  • Analytical
    RP-18e
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50550.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50329.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,60 mm
    100 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 440,00 Kč
    22 440,00 Kč
    1 * 1 KS
       22 440,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50578.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 910,00 Kč
    24 910,00 Kč
    1 * 1 KS
       24 910,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50579.0001
  • Analytical
    RP-8
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50580.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,60 mm
    100 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 19 960,00 Kč
    19 960,00 Kč
    1 * 1 KS
       19 960,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50581.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,60 mm
    150 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 24 910,00 Kč
    24 910,00 Kč
    1 * 1 KS
       24 910,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50582.0001
  • Analytical
    RP-8e
    100 Å
    5 µm
    4,60 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 27 160,00 Kč
    27 160,00 Kč
    1 * 1 KS
       27 160,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50583.0001
  • Analytical
    Si
    100 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 25 740,00 Kč
    25 740,00 Kč
    1 * 1 KS
       25 740,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50316.0001
LiChrospher® PAH LiChroCART® cartridges
Informace o produktu
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Dostupnost
    Price
  • Analytical
    PAH
    150 Å
    5 µm
    3,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 30 110,00 Kč
    30 110,00 Kč
    1 * 1 KS
       30 110,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50156.0001
  • Analytical
    PAH
    150 Å
    5 µm
    4,00 mm
    4 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 10 KS
    1 * 10 KS - 14 760,00 Kč
    14 760,00 Kč
    1 * 10 KS
       14 760,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50148.0001
  • Analytical
    PAH
    150 Å
    5 µm
    4,00 mm
    250 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 32 350,00 Kč
    32 350,00 Kč
    1 * 1 KS
       32 350,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.50149.0001