Shoe covers, Secutex® pro, PP/PE
These shoe covers protect against aggressive chemicals and other substances.
- Lint-free, resistant to abrasion, tear and water
- Permeable to air and water vapour ensures breathability
- Anti-static
*One size fits all.
- Catalog No:
- ZVGA30250-90
- ZVGA30254-90
- Garment application:
- Chemical protection, Laboratory
- Chemical protection, Laboratory
Frequently Bought Together