VWR®, Bouffant Caps, PP
Dodavatel: VWR Collection
Certifikáty
O tomto produktu
Bouffant caps made from non woven PP.
- Latex-free
- Elasticated band
Weight: 12 g/m²
Packaging: Supplied in boxes or bags.
Specifikace
- Catalog No:
- 113-8288
- 113-8289
- 113-8241
- 113-8243
- 113-8244
- 113-8245
- 113-8246
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
Specifikace
Frequently Bought Together