Burette clamps
Supplier: BOCHEM
Jump to:
About this item
Stainless steel. For simple and secure fixing of burettes.
- With bosshead for fixing to retort stand
- Clamping range 0 to 20 mm
- Jaws are covered with a silicone coating to protect the burettes
2 Options Available Below
Product Details & Documents
Stainless steel. For simple and secure fixing of burettes.
- With bosshead for fixing to retort stand
- Clamping range 0 to 20 mm
- Jaws are covered with a silicone coating to protect the burettes
Product Family Options
Product Information