ContactUsLinkComponent
Accessories for inoculating loops and needles
Accessories for inoculating loops and needles
Catalog # 631-2240
undefined
Accessories for inoculating loops and needles
Catalog # 631-2240
Specifications
  • Description:
    Stand for inoculating loops and needles
  • Pk:
    1
Specifications
About this item
PP. For storage of 6 needles and inoculation loops. Ø 120 mm.
  • Round
  • With 2×6 holes for Ø 7 and 9 mm
  • With central hole for loop holders