Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Cell scrapers, Nunc™
Cell scrapers, Nunc™
Cell scrapers, Nunc™
Cell scrapers, Nunc™
Cell scrapers for cell harvesting are available in two lengths, with adjustable blade for optimal application flexibility. Non-pyrogenic.
 : Supplied individually wrapped, one per sleeve.
  • Catalog No:
  • 734-2132
  • 734-2133
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Certifikát ochrany životního prostředí:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Product Family Options
  • Popis
    Celková délka
  • For use with 25 - 80 cm² flasks, blade offset 7.5 mm, blade width 15.5 mm
    230 mm
    1 * 250 KS
    1 * 250 KS - 36 130,00 Kč
    36 130,00 Kč
    1 * 250 KS
       36 130,00 Kč
      Low Manufacturing Impact
    Specifications
    Cat. No.
    734-2132
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • For use with 75 - 175 cm² flasks, blade offset 16 mm, blade width 17.5 mm
    320 mm
    1 * 250 KS
    1 * 250 KS - 41 310,00 Kč
    41 310,00 Kč
    1 * 250 KS
       41 310,00 Kč
      Low Manufacturing Impact
    Specifications
    Cat. No.
    734-2133
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact