Order Entry
Czech Republic
ContactUsLinkComponent
 
Cell counter cup, Coulter®
  720-0812
 :  KARTELL
undefined
Cell counter cup, Coulter®
  720-0812
 :  KARTELL

 

  • Description:
    Cell counter cups (Ø×H: 30×56 mm)
  • Pk:
    200

 

 

Polystyrene cup with polyethylene lid graduated at 10 and 20 ml.

  • Graduated at 10 and 20 ml

Ø×H: 30×56 mm

 : Cups and lids are packaged separately.