ContactUsLinkComponent
SP Bel-Art 'The Collector™' Blood Collecting Tray Set, Bel-Art Products, a part of SP
SP Bel-Art 'The Collector™' Blood Collecting Tray Set, Bel-Art Products, a part of SP
Catalog # 129-1015
undefined
SP Bel-Art 'The Collector™' Blood Collecting Tray Set, Bel-Art Products, a part of SP
Catalog # 129-1015
Specifications
  • Description:
    "The Collector™"
  • W×D×H:
    356×292×51 mm
  • Pk:
    1
Specifications
About this item
PP, white.
  • With two 40-place test tube racks (tubes up to 16 mm Ext.Ø.)
  • With one 18-place slide holder
  • With large requisition section
  • Adjustable partitions for syringes, needles, cotton etc.
"The Collector" is a versatile carrying tray particularly suited for use in clinical laboratories. The tray will not sag even when fully loaded. It is steam autoclavable at 121 °C. The handle is an end-mounted, detachable loop (178 mm high).