ContactUsLinkComponent
Dispenser for Disposable Items
Dispenser for Disposable Items
Catalog # 113-0031
Supplier:  AK
undefined
Dispenser for Disposable Items
Catalog # 113-0031
Supplier:  AK
Specifications
  • Description:
    Dual compartment dispenser
  • W×D×H:
    508×305×457 mm
  • Pk:
    1
Specifications
About this item
PETG.
For storing and dispensing disposable items such as overshoes, gloves or hoods and many other laboratory utensils.
  • Two compartments with separate hinged lids for simple filling and easy removal
  • Easy to clean
  • Robust, stable bench model