Bottles, amber, wide neck, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, amber with amber PP screw cap.
- Good chemical resistance
- Heat resistant up to 120 °C
- Leakproof
Amber coloured to protect light-sensitive materials. Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids.
Specifications
- Catalog No:
- 215-7235
- 215-7236
- 215-7233
- 215-7234
- 215-7231
- 215-7232
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement