Bottles, wide neck, with flared rim, DURAN®
DURAN®, borosilicate glass 3.3, clear, ungrounded neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Glass Type I according to USP, EP and JP
Round, with printed volumes and white labelling area.
: DIN 12038.
Frequently Bought Together