Bottles, narrow neck, square, HDPE, with screw cap, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
HDPE translucent, with LDPE screw cap.
- Square cross section bottles saves on storage space
- Bottles are available capped (assembled) or uncapped (unassembled)
A range of square, narrow mouth bottles. Ideal for use in the laboratory, production plant or in the field for storage of liquids, solids and powders.
Frequently Bought Together