ContactUsLinkComponent
Dropping bottles, FEP, Nalgene®
Dropping bottles, FEP, Nalgene®
Catalog # 215-7861
   Low Manufacturing Impact
undefined
Dropping bottles, FEP, Nalgene®
Catalog # 215-7861
   Low Manufacturing Impact
Specifications
  • Capacity:
    30 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
PTFE/FEP, with ETFE drip lock and cap.
  • Excellent chemical resistance
  • Heat resistant up to 150 °C, autoclavable
  • Attached sealing cap cannot be lost
Suitable for drop-dispensing practically any chemical.