ContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®
  215-1065
   Low Manufacturing Impact
undefined
Dropping bottles, LDPE, Nalgene®
  215-1065
   Low Manufacturing Impact
  • Capacity:
    15 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    25
LDPE, white with colour-coded (red, green, blue, white and yellow) PP screw caps and LDPE dosing tip.
  • Good chemical resistance, heat resistant up to 80 °C
  • Drop control tip snaps securely into place. Drops are dispensed individually
  • White material, ideal for UV light-sensitive products, leakproof
  • Delivers 40 µl drops (based on water; viscosity affects drop size)
Reagent identification is very easy with the varied colours of the screw caps. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.