ContactUsLinkComponent
Beakers, low form
Beakers, low form
Catalog # 213-0265
Supplier:  NICKEL ELECTRO
Supplier #: 207
undefined
Beakers, low form
Catalog # 213-0265
Supplier:  NICKEL ELECTRO
Supplier #: 207
Log in for availability
Select an Option
125 mm
Select an Option
2000 ml
Select an Option
180 mm
Select an Option
07
Select an Option
1
About this item
Stainless steel, rust-proof, with spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together