Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Non Hazardous Waste Bags
VWR®, Non Hazardous Waste Bags
Catalog # 129-0022
Supplier:  VWR Collection
undefined
VWR®, Non Hazardous Waste Bags
Catalog # 129-0022
Supplier:  VWR Collection

Specifications

  • Capacity:
    80 L
  • L×W:
    889×635 mm
  • Material:
    PP
  • Seq:
    04
  • Pk:
    200

Specifications

About this item

Sturdy bags, ideal for disposing of non hazardous waste that must first be autoclaved. In PP, 50 µm thick, transparent, with labelling area.

  • Autoclavable up to 121 °C
  • Highly resistant to tears and impact
  • Specially welded seams reduce risk of leakage

The bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709-98.

Certifications: Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709-98.