



Boron trifluoride - methanol complex 50% in methanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:373-57-9
:16045-88-8
Formula:CH₄BF₃O
MDL Number:MFCD00071635
:99.85 g/mol
:M-Clarity™ quality level = MQ200
EINECS:206-766-4
:GHS02
:GHS06
:GHS05
:GHS08
Frequently Bought Together