Order Entry
Czech Republic
ContactUsLinkComponent
Boron trifluoride - methanol complex 12% 1.5 M in methanol
Flammable
Toxic
Corrosive
Health
Boron trifluoride - methanol complex 12% 1.5 M in methanol
Certificates
undefined
Flammable
Toxic
Corrosive
Health
Boron trifluoride - methanol complex 12% 1.5 M in methanol
Certificates

Specification Test Results

  • Appearance:
    Colorless to amber Solution
  • Titration with NaOH:
    10 to 15 % w/v
  • Refractive index:
    1.5250 to 1.5300 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together