
Sodium diphenylamine-4-sulphonate, pure indicator grade, redox indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6152-67-6
:
Formula:C₁₂H₁₀NNaO₃S
MDL Number:MFCD00036468
:271.27 g/mol
Merck Index:13,07397
EINECS:228-165-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to beige to light grey Crystalline powder
- Infrared spectrum:Conforms
- Sensitivity:(as indicator) Passes test
Specification Test Results
Frequently Bought Together