Order Entry
Czech Republic
ContactUsLinkComponent
 
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™, Nalgene®
  513-1275
   Low Manufacturing Impact
undefined
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™, Nalgene®
  513-1275
   Low Manufacturing Impact

 

  • Membrane:
    PES
  • Pore size:
    0,45 µm
  • Capacity:
    115 ml
  • Description:
    Complete unit
  • Seq:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
  • Membrane Ø:
    50 mm
  • Pk:
    12

 

 

Rapid-Flow™ disposable filter units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination.

  • Color coded according to membrane

    Fastest flow rate and lowest clogging of comparable alternatives

    0.1 µm PES available for mycoplasma protection

    0.2 µm PES devices are stem cell and mouse embryonic assay (MEA) tested

Ideal for the filtration of cell culture media, serum, additives and buffers.

Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.

Available with Fit for Purpose Sterile documentation.