Tanks, cylindrical, with stopcock and cover, Nalgene®
These heavy-duty cylindrical LLDPE tanks with cover and spigot can withstand most acid, alcohols and bases. Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
- Economical
- Impact- and abrasion-resistant
- Not autoclavable
- Catalog No:
- NALG11102-0055
- NALG11102-0015
- NALG11102-0007
- NALG11102-0030
- NALG11102-0010
- NALG11102-0005
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together