

Tritolyl phosphate (mixture of isomers), Sigma-Aldrich®
Catalog # SIAL268917-500ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:368.37 g/mol
Merck Index:12,09892
Boiling Pt:420 °C (1013 hPa)
Melting Pt:–40 °C
Density:1.16 g/cm³ (20 °C)
EINECS:215-548-8
UN:2810
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:GHS07
:GHS09
CAS Number:1330-78-5
Formula:C₂₁H₂₁O₄P
MDL Number:MFCD02102116