

o,o,o-Tritolyl phosphate 96%
:78-30-8
:
:C₂₁H₂₁O₄P
:MFCD00041906
:368.37 g/mol
:12,09893
:410 °C (760 mmHg)
:–25 °C
:1.2 g/cm³
:225 °C
:Ambient
:201-103-5
:2574
:6.1,II
:GHS08
:GHS09
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5580 to 1.5610 (20°C, 589 nm)
- GC:≥95 %
Frequently Bought Together