2-Acetylanthracene, Sigma-Aldrich®
Catalog # SIAL566780-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:220.27 g/mol
CAS Number:10210-32-9
Formula:C₁₆H₁₂O
MDL Number:MFCD00156727