Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Light Green SF Yellowish
Light green SF yellowish, high purity
Light green SF yellowish, high purity
0394-25G
:
VWR Chemicals
:
Light green SF yellowish, high purity
0394-25G
:
VWR Chemicals
:
Pack type:
Bottle, amber
Pk:
25 g
Seq:
3308
:
:
792.86 g/mol
:
13,05506
:
288−289 °C
:
288…289 °C
:
Ambient
:
225-906-5
:
5141-20-8
:
C₃₇H₃₄N₂Na₂O₉S₃
:
MFCD00012121