Order Entry
Czech Republic
ContactUsLinkComponent
Titriplex® VI (EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)) analytical reagent, Supelco®
Titriplex® VI (EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)) analytical reagent, Supelco®
Supplier:  Merck
Certificates
Titriplex® VI (EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)) analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    240 - 245 °C decomposes
  • Molar Mass:
    380.35 g/mol
  • Bulk Density:
    450 kg/m3

Specification Test Results

Related Information

Frequently Bought Together