Order Entry
Czech Republic
ContactUsLinkComponent
Congo red 0.1% in ethanol 15% (v/v) Reag. Ph. Eur. 1022001
Congo red 0.1% in ethanol 15% (v/v) Reag. Ph. Eur. 1022001
  87805.180
 :  
undefined
Congo red 0.1% in ethanol 15% (v/v) Reag. Ph. Eur. 1022001
  87805.180
 :  

 

  • Pk:
    100 mL
  • Seq:
    2440

 

 

 :
 :696.67 g/mol
 :Ambient
 :209-358-4
 :573-58-0
 :C₃₂H₂₂N₆Na₂O₆S₂
 :MFCD00004028