
Methyl blue, high purity biological stain
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:28983-56-4
:
Formula:C₃₇H₂₇N₃Na₂O₉S₃
MDL Number:MFCD00058509
:799.81 g/mol
EINECS:249-113-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown Glistening crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:≤10 %
- UV Lambda max:594 to 610 nm (in 0.005M HCl)
- Absorption Ratio P-15/P+15:0.96 to 1.09
Specification Test Results
Frequently Bought Together