VWR® Desiccator Plates
Supplier: VWR Collection
Certificates
About this item
Porcelain desiccator plates. Central hole Ø 20 mm, filter holes Ø approximately 5 mm. Suitable for use with any desiccators with corresponding diameter.
Certifications: DIN 12911
Related Information
VWRI467-0102_TDS_EN
Technický list
VWR® for rotary evaporation VWR® Glassware Solutions
Produktová literatura
Frequently Bought Together