Order Entry
Czech Republic
ContactUsLinkComponent
 
Human/Mouse Recombinant TGF-β 3 (Animal free) (from E. coli)
 
Human/Mouse Recombinant TGF-β 3 (Animal free) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
  • Storage conditions:
    Store at 4 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Form:
    Liquid
  • Gene ID:
    P10600/P17125
  • Amino acid number:
    113/226
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein/peptide name:
    TGF-β 3
  • Purity:
    ≥95%
  • Molecular weight:
    12,9/25,7 kDa
  • Sequence:
    MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/ml
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    Ice pack
  • Tested applications:
    Bioassay
  • Purification:
    Reducing and non-reducing SDS-PAGE analysis

 

Frequently Bought Together