Order Entry
Czech Republic
ContactUsLinkComponent
Mouse Recombinant Neural SCE PB (from E. coli)
Mouse Recombinant Neural SCE PB (from E. coli)
Catalog # SHBTPB-500-18
New Product
CAS Number:  
Mouse Recombinant Neural SCE PB (from E. coli)
Catalog # SHBTPB-500-18
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 KIT
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    0075
  • Storage conditions:
    At or below −20 °C
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Endotoxin level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

About this item

IL-2C126S, an immune-regulating cytokine originating from lymphocytes, operates via the IL-2R receptor to trigger the expansion of activated T cells and encourage T cell maturation. IL-2 mediates its action by binding to IL-2 receptors (IL-2R), consisting of either trimeric receptors made of IL-2Rα (CD25), IL-2Rβ (CD122), and IL-2Rγ (γc, CD132) chains or dimeric βγ IL-2Rs.

  • High quality
  • Low endotoxin
  • Affordable

This cytokine also facilitates the growth and differentiation of B cells, natural killer (NK) cells, monocytes, and macrophages. Moreover, it plays a role in promoting the proliferation and differentiation of CAR T cells, T cells, and NK cells.In addition, IL-2 stimulates the activation and amplification of T cells and NK cells derived from peripheral blood.

Caution: For research use only.