Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant PDGF-BB (CTGrade GMP) (from E. coli)
Human Recombinant PDGF-BB (CTGrade GMP) (from E. coli)
Certificates
Human Recombinant PDGF-BB (CTGrade GMP) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.01
  • Biological activity:
    Bioactivity assay: Proliferation of NR6R-3T3, ED50 ≤5 ng/ml, bioactivity ≥1,6 x10³ IU/µg
  • Gene ID:
    P01127
  • Reconstitution instructions:
    Sterile water at 0,1 - 0,5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    ODGF|GDGF
  • Purity:
    ≥98%
  • Molecular weight:
    12,4/24,9 kDa
  • Sequence:
    MSLGSLTIAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRPTQVQLRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETVAAARPVT
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together