Human Glioma Culture Media Recombinant Protein Bundle (rh EGF, rh FGF-basic 154, rh PDGF-AA, rh PDGF-BB) (from E. coli)
Catalog # SHBTPB-500-19
Supplier: Shenandoah Biotechnology
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 KIT
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:0075
- Storage conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:Human glioma culture PB
- Endotoxin level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
About this item
Cytokines for cell culture of human DIPG and other glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Diffuse Intrinsic Pontine Glioma (DIPG) is a pediatric brain tumor that originates in the brainstem. Cells isolated from these childhood tumors are successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).
- High Quality
- Low Endotoxin
- Affordable
Host cell protein: ≤30 ng/ml
Host cell DNA: ≤0.01 ng/ml
Sterility Testing: No Growth
Mycoplasma: Negative
Delivery information: Bundle includes: 2×100 μg of Recombinant Human EGF, 500 μg of Recombinant Human FGF-basic 154 / FGF-2, 100 μg of Recombinant Human PDGF-AA, 100 μg of Recombinant Human PDGF-BB.
Caution: For research use only.