Order Entry
Czech Republic
ContactUsLinkComponent
Mouse T Regulatory Cell Polarizing Recombinant Protein Bundle (rm IL-2, rh TGF-β 1) (from E. coli)
Mouse T Regulatory Cell Polarizing Recombinant Protein Bundle (rm IL-2, rh TGF-β 1) (from E. coli)
Catalog # SHBTPB-500-10
CAS Number:  
Mouse T Regulatory Cell Polarizing Recombinant Protein Bundle (rm IL-2, rh TGF-β 1) (from E. coli)
Catalog # SHBTPB-500-10
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 KIT
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    0075
  • Storage conditions:
    At or below –20 °C
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein/peptide name:
    Mouse T reg cell polarizing PB
  • Endotoxin level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

About this item

Cytokines for polarizing T cells into T regulatory cells. T regulatory cells (Tregs) are a subset of T cells that suppress immune function. They are characterized by the expression of CD4, CD25 and Foxp3 and are known to produce immunosuppressive cytokines TGF-β 1 and IL-10. Tregs can be cultured from naive CD4+ T cells by culturing them in the presence of IL-2 and TGF-β 1 with CD3 and CD28 stimulation. In some cases cells are also cultured with neutralizing antibodies to IL-4, IFNγ and IL-12 and/or retinoic acid.

  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.