Order Entry
Czech Republic
ContactUsLinkComponent
Mouse Monocyte-Derived Dendritic Cell Differentiation Recombinant Protein Bundle (rm GM-CSF/rm IL-4) (from E. coli)
Mouse Monocyte-Derived Dendritic Cell Differentiation Recombinant Protein Bundle (rm GM-CSF/rm IL-4) (from E. coli)
Catalog # SHBTPB-500-02
New Product
Číslo CAS:  
Mouse Monocyte-Derived Dendritic Cell Differentiation Recombinant Protein Bundle (rm GM-CSF/rm IL-4) (from E. coli)
Catalog # SHBTPB-500-02
New Product
Číslo CAS:  

Specifikace

  • Pack type:
    Vial
  • Pk:
    1 KIT
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    0075
  • Storage conditions:
    At or below –20 °C
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein/peptide name:
    Mouse monocyte/DC PB
  • Endotoxin level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifikace

O tomto produktu

Cytokines for the differentation of mouse monycyte-derived dendritic cells. Dendritic cells are an important type of immune cell that function to activate T and B cells via their role as an antigen presenting cells. Monocytes isolated from bone marrow can be differentiated to immature dendritic cells (iDCs) through culturing in the presence of interleukin 4 (IL-4) and granulocyte-macrophage colony stimulating factor (GM-CSF). The characteristics of monocyte-derived iDCs are detectable after culture with IL-4/GM-CSF and include: the down regulation of CD14 surface expression, an increase in dextran uptake and an increased response to MIP-1α. Subsequent culturing with pro-inflammatory cytokines (such as TNF-α, IL-1β, and IL-6) can further differentiate the iDCs into mature dendritic cells (mDCs) that have full T cell stimulating capacity. This bundle includes 1x 100 μg of Recombinant Mouse IL-4 and 1x 100 μg of Recombinant Mouse GM-CSF.

  • High Quality
  • Low Endotoxin
  • Affordable

This bundle includes 1×100 μg of Recombinant Mouse IL-4 and 1×100 μg of Recombinant Mouse GM-CSF. Specific concentrations of each chemokine are determined by the end user based on their application and culture conditions.

Delivery: Bundle includes: 100 μg of recombinant mouse IL-4; 100 μg of recombinant mouse GM-CSF.
Caution: For research use only.