Millex® Syringe Filter Units, Hydrophobic PTFE Membrane
New Product
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
- Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.
Frequently Bought Together