ContactUsLinkComponent
Opticap® XL Filter
Opticap® XL Filter
Catalog # MILY0PTIA04NN1
Supplier:  MILLIPORE
New Product
Opticap® XL Filter
Catalog # MILY0PTIA04NN1
Supplier:  MILLIPORE
New Product
Specifications
  • Description:
    Opticap® XL filter, sterile, Durapore® membrane, 0,22 μm pore size
  • Environmentally Preferable:
Specifications
About this item
The Opticap® filter is a pharmaceutical-grade sterilizing filter designed to remove bacteria and particulates from pure water before it enters the analyser. Placed in the distribution loop, the loop panel with Opticap® XL Filter may be wall-mounted or standalone.
The filter is compatible with Milli-Q® CLX 7000 series water purification systems.
  • Produces bacteria- and particulate-free Type 2 pure water
  • 0,22 μm Durapore® membrane for bacteria removal
  • Ensure compliance with CLRW-quality specifications
  • Replace every six months for optimal results
Application:
The Opticap® XL filter delivers bacteria- and particulate-free purified water for critical research applications.