Multiscreen® Filter Plates, 96-Well, Hydrophilic PVDF Membrane
Supplier: MILLIPORE
New Product
Certificates
About this item
MultiScreen® HTS HV Filter Plate, 0,45 μm, opaque, non-sterile has been used in the maltose-binding protein (MBP)-E6 and E6 association protein (AP) binding assay.
- High throughput screening
- Rigid sidewalls
- Barcode-labeling capability
- Design that prevents cross-well contamination
L×W×H: 128×85,5×14,4 mm
Filtration area: 0.28 cm²
Binding type: Low binding surface
Certifications: The American National Standards Institute (ANSI) and the Society of Biomolecular Screening (SBS) standards compliance.
Frequently Bought Together