Millex® Samplicity Syringe Filters
Dodavatel: MILLIPORE
New Product
Certifikáty
O tomto produktu
Millex® Filters for the Samplicity® G2 System
- Samplicity® G2 filtration system enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials
- Millex® Filters for the Samplicity® G2 system provide adaptor funnels and Millex® 33 mm filters
- Samplicity® G2 system has lower ergonomic impact than manual syringe filtration
- Available in 0,20 µm and 0,45 µm pore sizes with hydrophilic PTFE, Durapore® PVDF, Millipore Express® PLUS PES, or nylon membranes
Applications: HPLC; filtration of viscous or particulate laden samples for HPLC.
Frequently Bought Together