
Oxalic acid
Catalog # CMPLSB65562.250MG
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
CAS Number:
Specifications
- Pk:250 mg
Specifications
About this item
:
:90.04 g/mol
Boiling Pt:>100 °C
Melting Pt:189,5 °C
Density:1,901 g/cm³
EINECS:205-634-3
:GHS07
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573