Anti-Fractin Rabbit Polyclonal Antibody
NOVUNBP2-29504
New Product
- Antibody type:Primary
- Antigen name:Fractin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:Neat whole antisera
- Storage temperature:Store at −20 °C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to amino acid residues from the C terminal region of the 32 kDa actin fragment
- Pack type:Vial
- Pk:0,1 mL
Type: Primary
Antigen: Fractin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human