Order Entry
Czech Republic
ContactUsLinkComponent
Water, Biotech Reagent for injection (WFI), J.T.Baker®
Water, Biotech Reagent for injection (WFI), J.T.Baker®
Catalog # 2910-97
Supplier:  Avantor
New Product
CAS Number:  
undefined
Water, Biotech Reagent for injection (WFI), J.T.Baker®
Catalog # 2910-97
Supplier:  Avantor
New Product
CAS Number:  

Specifications

  • Pack type:
    Single-use bag in 219 L open head drum
  • Pk:
    20 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Appearance (Clear & Colorless liquid):
    Passes Test
  • Bacterial Endotoxins (EU/mL):
    <=0.25 EU/mL
  • Bioburden:
    <=10 CFU/mL
  • pH @ 25° C:
    5 to 7
  • Total Organic Carbon (TOC) recorded from recirculation loop:
    <200 ppm
  • Nitrates (NO3):
    <0.2 ppm

Specification Test Results

About this item

 :
 :18.02 g/mol
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
 :Filtered through a 0.2 µm filter.
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332