Order Entry
Czech Republic
ContactUsLinkComponent
Titan yellow 0,05% in aqueous solution Reag. Ph. Eur. 1090902
Titan yellow 0,05% in aqueous solution Reag. Ph. Eur. 1090902
Catalog # 87955.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Titan yellow 0,05% in aqueous solution Reag. Ph. Eur. 1090902
Catalog # 87955.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :695.73 g/mol
EINECS:217-377-4
CAS Number:1829-00-1
Formula:C₂₈H₁₉N₅Na₂O₆S₄
MDL Number:MFCD00013091