Order Entry
Czech Republic
ContactUsLinkComponent
Potassium heptaiodobismuthate 0,5% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070603
Potassium heptaiodobismuthate 0,5% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070603
Catalog # 87915.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium heptaiodobismuthate 0,5% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070603
Catalog # 87915.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :1253.71 g/mol
EINECS:255-595-1
CAS Number:41944-01-8
Formula:BiI₇K₄