Order Entry
Czech Republic
ContactUsLinkComponent
Potassium heptaiodobismuthate 8% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070602
Potassium heptaiodobismuthate 8% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070602
Catalog # 87914.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium heptaiodobismuthate 8% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070602
Catalog # 87914.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :1253.71 g/mol
EINECS:255-595-1
CAS Number:41944-01-8
Formula:BiI₇K₄